The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304725.1 | NP_001291654.1 | 4 - 5 | 609 | 77 |
NM_017655.5 | NP_060125.4 | 4 - 5 | 906 | 77 | |
GenBank mRNA | AB073737.1 | - | 4 - 5 | 722 | 77 |
AK000082.1 | - | 3 - 4 | 465 | 77 | |
AK225941.1 | - | 5 - 6 | 745 | 77 | |
AK307422.1 | - | 4 - 5 | 609 | 77 | |
BC036075.1 | - | 5 - 6 | 906 | 77 |