Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FAM208B
- Entrez Gene ID
-
54906
- Gene Name
- family with sequence similarity 208 member B
- Gene Aliases
-
C10orf18, bA318E3.2
- Chromosome Location
-
Chr.10: 5684759 - 5763740 on Build GRCh38
View Full Product Details