The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024848.2 | NP_079124.1 | 11 - 12 | 1395 | 63 |
XM_005244798.2 | XP_005244855.1 | 10 - 11 | 1259 | 63 | |
XM_011542170.2 | XP_011540472.1 | 11 - 12 | 1395 | 63 | |
XM_011542172.2 | XP_011540474.1 | 10 - 11 | 1259 | 63 | |
XM_011542173.2 | XP_011540475.1 | 11 - 12 | 1395 | 63 | |
XM_011542174.2 | XP_011540476.1 | 11 - 12 | 1395 | 63 | |
XM_011542176.1 | XP_011540478.1 | 9 - 10 | 1104 | 63 | |
GenBank mRNA | AK024003.1 | - | 11 - 12 | 1198 | 63 |
AK310323.1 | - | 10 - 11 | 1259 | 63 |