The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001280797.1 | NP_001267726.1 | 8 - 9 | 1092 | 63 |
NM_001280798.1 | NP_001267727.1 | 6 - 7 | 870 | 63 | |
NM_013267.3 | NP_037399.2 | 9 - 10 | 1222 | 63 | |
XM_005268797.1 | XP_005268854.1 | 9 - 10 | 849 | 63 | |
XM_017019180.1 | XP_016874669.1 | 8 - 9 | 719 | 63 | |
GenBank mRNA | AF110330.1 | - | 9 - 10 | 977 | 63 |
AF223944.1 | - | 9 - 10 | 1149 | 63 | |
AK000467.1 | - | 9 - 10 | 969 | 63 | |
BC025661.1 | - | 8 - 9 | 807 | 63 | |
DA060302.1 | - | 6 - 7 | 454 | 63 | |
HM775422.1 | - | 8 - 9 | 841 | 63 | |
HM775423.1 | - | 7 - 8 | 711 | 63 |