Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DSC1
Entrez Gene ID
1823
Gene Name
desmocollin 1
Gene Aliases
CDHF1, DG2/DG3
Chromosome Location
Chr.18: 31129236 - 31162856 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004948.3 NP_004939.1 12 - 13 2138 113
NM_024421.2 NP_077739.1 12 - 13 2138 113
GenBank mRNA X72925.1 - 12 - 13 2138 113
Z34522.1 - 12 - 13 2066 113

View Full Product Details