The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004948.3 | NP_004939.1 | 4 - 5 | 731 | 63 |
NM_024421.2 | NP_077739.1 | 4 - 5 | 731 | 63 | |
GenBank mRNA | X72925.1 | - | 4 - 5 | 731 | 63 |
Z34522.1 | - | 4 - 5 | 659 | 63 |