The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258308.1 | NP_001245237.1 | 4 - 5 | 369 | 114 |
NM_001258309.1 | NP_001245238.1 | 4 - 5 | 369 | 114 | |
XM_005253691.1 | XP_005253748.1 | 4 - 5 | 402 | 114 | |
GenBank mRNA | AK307582.1 | - | 4 - 5 | 351 | 114 |
AU131299.1 | - | 4 - 5 | 351 | 114 | |
BC000656.2 | - | 4 - 5 | 290 | 114 | |
BC106072.1 | - | 4 - 5 | 315 | 114 | |
BC128183.1 | - | 4 - 5 | 344 | 114 | |
BC128184.1 | - | 4 - 5 | 344 | 114 | |
DC316656.1 | - | 4 - 5 | 369 | 114 | |
M32110.1 | - | 3 - 4 | 283 | 114 | |
X55504.1 | - | 3 - 4 | 239 | 114 |