The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001584.2 | NP_001575.1 | 5 - 6 | 889 | 79 |
XM_005253110.3 | XP_005253167.1 | 6 - 7 | 1562 | 79 | |
XM_005253111.2 | XP_005253168.1 | 6 - 7 | 1199 | 79 | |
XM_005253112.1 | XP_005253169.1 | 6 - 7 | 1027 | 79 | |
XM_011520346.1 | XP_011518648.1 | 7 - 8 | 1066 | 79 | |
XM_011520347.2 | XP_011518649.1 | 6 - 7 | 1478 | 79 | |
XM_017018231.1 | XP_016873720.1 | 7 - 8 | 1048 | 79 | |
XM_017018233.1 | XP_016873722.1 | 7 - 8 | 1585 | 79 | |
XM_017018234.1 | XP_016873723.1 | 7 - 8 | 1697 | 79 | |
XM_017018235.1 | XP_016873724.1 | 7 - 8 | 1660 | 79 | |
XM_017018236.1 | XP_016873725.1 | 5 - 6 | 1380 | 79 | |
XM_017018237.1 | XP_016873726.1 | 5 - 6 | 960 | 79 | |
GenBank mRNA | AK309809.1 | - | 7 - 8 | 1066 | 79 |
BC031582.1 | - | 5 - 6 | 905 | 79 | |
U57911.1 | - | 5 - 6 | 889 | 79 |