Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- USP54
- Entrez Gene ID
-
159195
- Gene Name
- ubiquitin specific peptidase 54
- Gene Aliases
-
C10orf29, bA137L10.3, bA137L10.4
- Chromosome Location
-
Chr.10: 73497538 - 73626844 on Build GRCh38
View Full Product Details