The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024593.1 | NP_001019764.1 | 4 - 5 | 568 | 62 |
GenBank mRNA | AK309870.1 | - | 5 - 6 | 624 | 62 |
BC094693.1 | - | 4 - 5 | 547 | 62 | |
BC109255.1 | - | 3 - 4 | 481 | 62 | |
BC109256.1 | - | 3 - 4 | 481 | 62 | |
CR936824.1 | - | 4 - 5 | 550 | 62 |