Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGTPBP1
Entrez Gene ID
23287
Gene Name
ATP/GTP binding protein 1
Gene Aliases
CCP1, NNA1
Chromosome Location
Chr.9: 85543799 - 85742029 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286715.1 NP_001273644.1 12 - 13 1523 94
NM_001286717.1 NP_001273646.1 12 - 13 1403 94
NM_015239.2 NP_056054.2 13 - 14 1468 94
XM_005251847.3 XP_005251904.1 13 - 14 1534 94
XM_005251848.2 XP_005251905.1 13 - 14 1712 94
XM_005251849.3 XP_005251906.1 12 - 13 1458 94
XM_011518418.2 XP_011516720.1 13 - 14 1534 94
XM_011518420.2 XP_011516722.1 3 - 4 4787 94
XM_011518421.2 XP_011516723.1 13 - 14 1534 94
XM_017014544.1 XP_016870033.1 13 - 14 2079 94
XM_017014545.1 XP_016870034.1 13 - 14 1414 94
XM_017014546.1 XP_016870035.1 12 - 13 1301 94
XM_017014547.1 XP_016870036.1 12 - 13 1458 94
XM_017014548.1 XP_016870037.1 12 - 13 1401 94
XM_017014549.1 XP_016870038.1 11 - 12 1420 94
XM_017014550.1 XP_016870039.1 13 - 14 1534 94
GenBank mRNA AB028958.2 - 13 - 14 1446 94
AK001544.1 - 14 - 15 1268 94
AK022562.1 - 1 - 2 111 94
AK023280.1 - 1 - 2 103 94
AK295304.1 - 9 - 10 962 94
AK299506.1 - 9 - 10 963 94
AL833359.1 - 1 - 2 60 94
BC060815.1 - 13 - 14 1433 94
BX648366.1 - 1 - 2 515 94

View Full Product Details