The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021939.3 | NP_068758.3 | 5 - 6 | 1242 | 85 |
XM_011525099.2 | XP_011523401.1 | 5 - 6 | 1690 | 85 | |
XM_011525100.2 | XP_011523402.1 | 4 - 5 | 817 | 85 | |
GenBank mRNA | AB045981.1 | - | 5 - 6 | 1003 | 85 |
AF337909.1 | - | 5 - 6 | 917 | 85 | |
AK025694.1 | - | 4 - 5 | 674 | 85 | |
AK025874.1 | - | 1 - 2 | 108 | 85 | |
AK092042.1 | - | 4 - 5 | 817 | 85 | |
AK092708.1 | - | 4 - 5 | 906 | 85 | |
AK172811.1 | - | 5 - 6 | 948 | 85 | |
AK296107.1 | - | 5 - 6 | 739 | 85 | |
AL832985.1 | - | 5 - 6 | 984 | 85 | |
BC016467.1 | - | 5 - 6 | 977 | 85 | |
BX537452.1 | - | 5 - 6 | 997 | 85 |