The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258205.1 | NP_001245134.1 | 9 - 10 | 1570 | 61 |
NM_145068.3 | NP_659505.1 | 9 - 10 | 1570 | 61 | |
GenBank mRNA | AF514998.1 | - | 9 - 10 | 1312 | 61 |
AJ487035.2 | - | 9 - 10 | 1570 | 61 | |
AK127726.1 | - | 3 - 4 | 522 | 61 | |
AK304768.1 | - | 1 - 2 | 237 | 61 | |
AY118267.1 | - | 9 - 10 | 1380 | 61 | |
AY118268.1 | - | 9 - 10 | 1380 | 61 | |
BC104866.1 | - | 9 - 10 | 1342 | 61 | |
BC104868.1 | - | 9 - 10 | 1342 | 61 | |
BC143294.1 | - | 10 - 11 | 1511 | 61 | |
BC143295.1 | - | 11 - 12 | 1588 | 61 | |
BC143299.1 | - | 10 - 11 | 1511 | 61 | |
BC143330.1 | - | 9 - 10 | 1342 | 61 | |
BC143333.1 | - | 10 - 11 | 1431 | 61 | |
BC143334.1 | - | 9 - 10 | 1424 | 61 | |
BX537539.1 | - | 3 - 4 | 569 | 61 | |
EU295940.1 | - | 8 - 9 | 1248 | 61 |