The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173517.4 | NP_775788.2 | 1 - 2 | 302 | 111 |
GenBank mRNA | AK289522.1 | - | 1 - 2 | 215 | 111 |
AY423045.1 | - | 1 - 2 | 221 | 111 | |
BC027734.1 | - | 1 - 2 | 302 | 111 | |
BX647631.1 | - | 1 - 2 | 235 | 111 |