Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRERF1
Entrez Gene ID
55809
Gene Name
transcriptional regulating factor 1
Gene Aliases
BCAR2, HSA277276, RAPA, TREP132, TReP-132, dJ139D8.5
Chromosome Location
Chr.6: 42224933 - 42452216 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297573.1 NP_001284502.1 14 - 15 3374 86
NM_033502.3 NP_277037.1 14 - 15 3314 86
XM_011514741.2 XP_011513043.1 15 - 16 4011 86
XM_011514742.2 XP_011513044.1 17 - 18 4187 86
XM_011514743.2 XP_011513045.1 16 - 17 4110 86
XM_011514744.1 XP_011513046.1 15 - 16 3619 86
XM_011514745.1 XP_011513047.1 15 - 16 3513 86
XM_017011034.1 XP_016866523.1 15 - 16 4061 86
XM_017011035.1 XP_016866524.1 15 - 16 3729 86
XM_017011036.1 XP_016866525.1 15 - 16 3548 86
XM_017011037.1 XP_016866526.1 15 - 16 3579 86
XM_017011038.1 XP_016866527.1 17 - 18 4201 86
XM_017011039.1 XP_016866528.1 15 - 16 3806 86
XM_017011040.1 XP_016866529.1 15 - 16 3498 86
XM_017011041.1 XP_016866530.1 15 - 16 3620 86
XM_017011042.1 XP_016866531.1 14 - 15 4723 86
XM_017011043.1 XP_016866532.1 14 - 15 5733 86
XM_017011044.1 XP_016866533.1 15 - 16 4010 86
XM_017011045.1 XP_016866534.1 15 - 16 3950 86
XM_017011046.1 XP_016866535.1 14 - 15 3792 86
XM_017011047.1 XP_016866536.1 15 - 16 3872 86
XM_017011048.1 XP_016866537.1 14 - 15 3792 86
XM_017011049.1 XP_016866538.1 14 - 15 3732 86
XM_017011050.1 XP_016866539.1 15 - 16 4011 86
XM_017011051.1 XP_016866540.1 14 - 15 3609 86
XM_017011052.1 XP_016866541.1 13 - 14 3483 86
XM_017011053.1 XP_016866542.1 15 - 16 3659 86
XM_017011054.1 XP_016866543.1 13 - 14 3483 86
XM_017011055.1 XP_016866544.1 14 - 15 2317 86
GenBank mRNA AF297872.1 - 14 - 15 3308 86
AJ277275.1 - 13 - 14 2849 86
AJ277276.1 - 13 - 14 2849 86
AK002096.1 - 1 - 2 48 86
AK127493.1 - 10 - 11 2788 86
AM404182.1 - 14 - 15 3374 86
AM404183.1 - 14 - 15 3314 86
AM404259.1 - 14 - 15 3314 86

View Full Product Details