Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STAB2
Entrez Gene ID
55576
Gene Name
stabilin 2
Gene Aliases
FEEL2, FELE-2, FELL2, FEX2, HARE, SCARH1
Chromosome Location
Chr.12: 103587250 - 103766724 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017564.9 NP_060034.9 42 - 43 4723 65
XM_011538537.2 XP_011536839.1 42 - 43 4764 65
XM_011538538.2 XP_011536840.1 42 - 43 4764 65
XM_011538539.2 XP_011536841.1 28 - 29 2967 65
XM_011538541.2 XP_011536843.1 20 - 21 2074 65
XM_011538542.2 XP_011536844.1 9 - 10 883 65
XM_017019585.1 XP_016875074.1 42 - 43 4764 65
GenBank mRNA AB052958.1 - 42 - 43 4719 65
AJ295695.1 - 42 - 43 4723 65
AK024503.1 - 10 - 11 1065 65
AK074051.1 - 20 - 21 2094 65
AL133021.1 - 2 - 3 205 65
AY227444.1 - 11 - 12 1132 65
AY311388.1 - 42 - 43 4741 65

View Full Product Details