The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017564.9 | NP_060034.9 | 51 - 52 | 5645 | 87 |
XM_011538537.2 | XP_011536839.1 | 51 - 52 | 5686 | 87 | |
XM_011538538.2 | XP_011536840.1 | 51 - 52 | 5686 | 87 | |
XM_011538539.2 | XP_011536841.1 | 37 - 38 | 3889 | 87 | |
XM_011538541.2 | XP_011536843.1 | 29 - 30 | 2996 | 87 | |
XM_011538542.2 | XP_011536844.1 | 18 - 19 | 1805 | 87 | |
XM_017019585.1 | XP_016875074.1 | 51 - 52 | 5686 | 87 | |
GenBank mRNA | AB052958.1 | - | 51 - 52 | 5641 | 87 |
AF160476.1 | - | 6 - 7 | 667 | 87 | |
AJ295695.1 | - | 51 - 52 | 5645 | 87 | |
AK024503.1 | - | 19 - 20 | 1987 | 87 | |
AK074051.1 | - | 29 - 30 | 3016 | 87 | |
AK160380.1 | - | 50 - 51 | 5363 | 87 | |
AL133021.1 | - | 11 - 12 | 1127 | 87 | |
AY227444.1 | - | 20 - 21 | 2054 | 87 | |
AY311388.1 | - | 51 - 52 | 5663 | 87 |