Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STAB2
Entrez Gene ID
55576
Gene Name
stabilin 2
Gene Aliases
FEEL2, FELE-2, FELL2, FEX2, HARE, SCARH1
Chromosome Location
Chr.12: 103587250 - 103766724 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017564.9 NP_060034.9 60 - 61 6767 92
XM_011538537.2 XP_011536839.1 60 - 61 6808 92
XM_011538538.2 XP_011536840.1 60 - 61 6808 92
XM_011538539.2 XP_011536841.1 46 - 47 5011 92
XM_011538541.2 XP_011536843.1 38 - 39 4118 92
XM_011538542.2 XP_011536844.1 27 - 28 2927 92
XM_017019585.1 XP_016875074.1 60 - 61 6808 92
GenBank mRNA AB052958.1 - 60 - 61 6763 92
AF160476.1 - 15 - 16 1789 92
AJ295695.1 - 60 - 61 6767 92
AK024503.1 - 28 - 29 3109 92
AK074051.1 - 38 - 39 4138 92
AK160380.1 - 59 - 60 6485 92
AL133021.1 - 20 - 21 2249 92
AY227444.1 - 29 - 30 3176 92
AY311388.1 - 60 - 61 6785 92

View Full Product Details