Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CARS
Entrez Gene ID
833
Gene Name
cysteinyl-tRNA synthetase
Gene Aliases
CARS1, CYSRS, MGC:11246
Chromosome Location
Chr.11: 3000922 - 3057451 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014437.2 NP_001014437.1 14 - 15 1710 112
NM_001194997.1 NP_001181926.1 14 - 15 1710 112
NM_001751.5 NP_001742.1 13 - 14 1461 112
NM_139273.3 NP_644802.1 13 - 14 1461 112
NR_036542.1 14 - 15 1757 112
XM_006718341.3 XP_006718404.1 13 - 14 1484 112
XM_017018389.1 XP_016873878.1 13 - 14 1484 112
XM_017018390.1 XP_016873879.1 14 - 15 1724 112
XM_017018391.1 XP_016873880.1 11 - 12 1708 112
XM_017018392.1 XP_016873881.1 13 - 14 1646 112
GenBank mRNA AF288206.1 - 13 - 14 1394 112
AF288207.1 - 13 - 14 1394 112
AK096313.1 - 14 - 15 1724 112
AK222614.1 - 13 - 14 1402 112
AK289432.1 - 13 - 14 1434 112
AK295371.1 - 13 - 14 1352 112
AK296763.1 - 14 - 15 1575 112
AK298514.1 - 14 - 15 1677 112
AK302644.1 - 14 - 15 1677 112
AK308005.1 - 14 - 15 1630 112
BC002880.2 - 13 - 14 1447 112
BT009913.1 - 13 - 14 1377 112
BX647906.1 - 14 - 15 1684 112
L06845.1 - 11 - 12 1248 112

View Full Product Details