Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARS
Entrez Gene ID
833
Gene Name
cysteinyl-tRNA synthetase
Gene Aliases
CARS1, CYSRS, MGC:11246
Chromosome Location
Chr.11: 3000922 - 3057451 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014437.2 NP_001014437.1 2 - 3 359 116
NM_001194997.1 NP_001181926.1 2 - 3 359 116
NR_036542.1 2 - 3 359 116
XM_017018390.1 XP_016873879.1 2 - 3 326 116
GenBank mRNA AK096313.1 - 2 - 3 326 116
AK298514.1 - 2 - 3 326 116
AK302644.1 - 2 - 3 326 116
AK308005.1 - 2 - 3 279 116
BX647906.1 - 2 - 3 333 116

View Full Product Details