Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARS
Entrez Gene ID
833
Gene Name
cysteinyl-tRNA synthetase
Gene Aliases
CARS1, CYSRS, MGC:11246
Chromosome Location
Chr.11: 3000922 - 3057451 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014437.2 NP_001014437.1 4 - 5 544 154
NM_001194997.1 NP_001181926.1 4 - 5 544 154
NM_001751.5 NP_001742.1 3 - 4 295 154
NM_139273.3 NP_644802.1 3 - 4 295 154
NR_036542.1 4 - 5 591 154
XM_006718341.3 XP_006718404.1 3 - 4 318 154
XM_017018389.1 XP_016873878.1 3 - 4 318 154
XM_017018390.1 XP_016873879.1 4 - 5 558 154
XM_017018392.1 XP_016873881.1 3 - 4 480 154
GenBank mRNA AB208814.1 - 3 - 4 271 154
AF288206.1 - 3 - 4 228 154
AF288207.1 - 3 - 4 228 154
AK096313.1 - 4 - 5 558 154
AK222614.1 - 3 - 4 236 154
AK289432.1 - 3 - 4 268 154
AK295371.1 - 3 - 4 186 154
AK296763.1 - 4 - 5 409 154
AK298514.1 - 4 - 5 511 154
AK302644.1 - 4 - 5 511 154
AK308005.1 - 4 - 5 464 154
AU133264.1 - 3 - 4 285 154
BC002880.2 - 3 - 4 281 154
BT009913.1 - 3 - 4 211 154
BX647906.1 - 4 - 5 518 154
DC376478.1 - 3 - 4 295 154
L06845.1 - 1 - 2 82 154

View Full Product Details