The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153046.2 | NP_694591.2 | 7 - 8 | 1037 | 88 |
XM_005267309.3 | XP_005267366.1 | 7 - 8 | 1174 | 88 | |
XM_006720019.3 | XP_006720082.1 | 7 - 8 | 1174 | 88 | |
XM_006720020.3 | XP_006720083.1 | 6 - 7 | 1093 | 88 | |
XM_011536397.2 | XP_011534699.1 | 7 - 8 | 1174 | 88 | |
XM_011536398.2 | XP_011534700.1 | 7 - 8 | 979 | 88 | |
XM_011536400.2 | XP_011534702.1 | 7 - 8 | 1174 | 88 | |
XM_011536401.2 | XP_011534703.1 | 7 - 8 | 912 | 88 | |
GenBank mRNA | AK125978.1 | - | 2 - 3 | 186 | 88 |
AL833915.1 | - | 4 - 5 | 427 | 88 | |
BC128057.1 | - | 2 - 3 | 176 | 88 | |
U56244.1 | - | 6 - 7 | 776 | 88 |