Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDRD9
Entrez Gene ID
122402
Gene Name
tudor domain containing 9
Gene Aliases
C14orf75, HIG-1, NET54
Chromosome Location
Chr.14: 103928343 - 104052667 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153046.2 NP_694591.2 9 - 10 1209 71
XM_005267309.3 XP_005267366.1 9 - 10 1346 71
XM_006720019.3 XP_006720082.1 9 - 10 1346 71
XM_006720020.3 XP_006720083.1 8 - 9 1265 71
XM_011536397.2 XP_011534699.1 9 - 10 1346 71
XM_011536398.2 XP_011534700.1 9 - 10 1151 71
XM_011536400.2 XP_011534702.1 9 - 10 1346 71
XM_011536401.2 XP_011534703.1 9 - 10 1084 71
XM_011536402.2 XP_011534704.1 2 - 3 182 71
XM_017020955.1 XP_016876444.1 8 - 9 1096 71
GenBank mRNA AK125978.1 - 4 - 5 358 71
AL833915.1 - 6 - 7 599 71
BC128057.1 - 4 - 5 348 71
BU430972.1 - 1 - 2 57 71
U56244.1 - 8 - 9 948 71

View Full Product Details