Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A19
Entrez Gene ID
60386
Gene Name
solute carrier family 25 member 19
Gene Aliases
DNC, MCPHA, MUP1, THMD3, THMD4, TPC
Chromosome Location
Chr.17: 75272980 - 75289957 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021734.4 NP_068380.3 1 - 2 92 111
XM_011525098.1 XP_011523400.1 1 - 2 600 111
GenBank mRNA DA605220.1 - 1 - 2 73 111

View Full Product Details