The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005862.2 | NP_001005862.1 | 9 - 10 | 1247 | 55 |
NM_001289936.1 | NP_001276865.1 | 10 - 11 | 1298 | 55 | |
NM_001289937.1 | NP_001276866.1 | 6 - 7 | 1022 | 55 | |
NM_001289938.1 | NP_001276867.1 | 9 - 10 | 1247 | 55 | |
NM_004448.3 | NP_004439.2 | 6 - 7 | 1022 | 55 | |
NR_110535.1 | 6 - 7 | 1085 | 55 | ||
GenBank mRNA | AF177761.2 | - | 6 - 7 | 773 | 55 |
AK131568.1 | - | 4 - 5 | 1453 | 55 | |
AK295195.1 | - | 10 - 11 | 1298 | 55 | |
AK296908.1 | - | 6 - 7 | 837 | 55 | |
AK313683.1 | - | 6 - 7 | 976 | 55 | |
BC080193.1 | - | 6 - 7 | 983 | 55 | |
BC110392.1 | - | 9 - 10 | 1212 | 55 | |
BC167147.1 | - | 6 - 7 | 764 | 55 | |
KJ534831.1 | - | 6 - 7 | 761 | 55 | |
KJ534964.1 | - | 6 - 7 | 761 | 55 | |
M11730.1 | - | 6 - 7 | 911 | 55 | |
X03363.1 | - | 6 - 7 | 935 | 55 |