The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284400.2 | NP_001271329.1 | 8 - 9 | 1105 | 77 |
NM_001321678.1 | NP_001308607.1 | 8 - 9 | 953 | 77 | |
GenBank mRNA | AY358082.1 | - | 8 - 9 | 962 | 77 |