The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278167.1 | NP_001265096.1 | 9 - 10 | 1361 | 96 |
NM_001278168.1 | NP_001265097.1 | 8 - 9 | 1130 | 96 | |
NM_001278169.1 | NP_001265098.1 | 9 - 10 | 1129 | 96 | |
NM_016082.4 | NP_057166.4 | 9 - 10 | 1213 | 96 | |
NM_016408.3 | NP_057492.2 | 9 - 10 | 1361 | 96 | |
XM_011528856.2 | XP_011527158.1 | 6 - 7 | 2097 | 96 | |
XM_011528857.2 | XP_011527159.1 | 8 - 9 | 973 | 96 | |
XM_011528858.1 | XP_011527160.1 | 8 - 9 | 1212 | 96 | |
XM_011528859.2 | XP_011527161.1 | 8 - 9 | 972 | 96 | |
XM_011528860.1 | XP_011527162.1 | 6 - 7 | 672 | 96 | |
XM_017027876.1 | XP_016883365.1 | 8 - 9 | 1111 | 96 | |
XM_017027877.1 | XP_016883366.1 | 3 - 4 | 425 | 96 | |
GenBank mRNA | AF152097.1 | - | 10 - 11 | 1339 | 96 |
AF161516.1 | - | 9 - 10 | 1309 | 96 | |
AK023992.1 | - | 9 - 10 | 1311 | 96 | |
AK027449.1 | - | 9 - 10 | 1161 | 96 | |
AK222745.1 | - | 9 - 10 | 1311 | 96 | |
AK292075.1 | - | 9 - 10 | 1079 | 96 | |
AK295342.1 | - | 9 - 10 | 1297 | 96 | |
AK314532.1 | - | 9 - 10 | 1311 | 96 | |
AY462283.1 | - | 9 - 10 | 1312 | 96 | |
BC001215.1 | - | 8 - 9 | 1059 | 96 | |
BC050706.1 | - | 9 - 10 | 1304 | 96 | |
BM792566.1 | - | 1 - 2 | 53 | 96 | |
CX785345.1 | - | 4 - 5 | 568 | 96 | |
KU178644.1 | - | 7 - 8 | 968 | 96 | |
KU178645.1 | - | 8 - 9 | 1199 | 96 |