Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMO3
Entrez Gene ID
55885
Gene Name
LIM domain only 3
Gene Aliases
RBTN3, RBTNL2, RHOM3, Rhom-3
Chromosome Location
Chr.12: 16548372 - 16610233 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001395.2 NP_001001395.1 1 - 2 273 77
NR_045014.1 1 - 2 273 77
GenBank mRNA AK095595.1 - 1 - 2 77
BC017777.1 - 1 - 2 77
BI553855.1 - 1 - 2 77
DA780743.1 - 1 - 2 77

View Full Product Details