Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR37
Entrez Gene ID
22884
Gene Name
WD repeat domain 37
Gene Aliases
-
Chromosome Location
Chr.10: 1056836 - 1132297 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014023.3 NP_054742.2 10 - 11 1137 84
GenBank mRNA AB023199.1 - 10 - 11 1108 84
AK128194.1 - 9 - 10 3949 84
AK292591.1 - 10 - 11 1103 84
AK307259.1 - 9 - 10 1030 84
AL136827.1 - 10 - 11 1115 84

View Full Product Details