Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR37
Entrez Gene ID
22884
Gene Name
WD repeat domain 37
Gene Aliases
-
Chromosome Location
Chr.10: 1056836 - 1132297 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014023.3 NP_054742.2 7 - 8 772 76
GenBank mRNA AB023199.1 - 7 - 8 743 76
AK128194.1 - 6 - 7 3581 76
AK292591.1 - 7 - 8 735 76
AK307259.1 - 6 - 7 665 76
AL136827.1 - 7 - 8 750 76
BC018044.1 - 7 - 8 801 76

View Full Product Details