Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR37
Entrez Gene ID
22884
Gene Name
WD repeat domain 37
Gene Aliases
-
Chromosome Location
Chr.10: 1056836 - 1132297 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK128194.1 - 8 - 9 3706 68
AK292591.1 - 9 - 10 860 68

View Full Product Details