Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN32
Entrez Gene ID
10077
Gene Name
tetraspanin 32
Gene Aliases
ART1, PHEMX, PHMX, TSSC6
Chromosome Location
Chr.11: 2299480 - 2318201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139022.2 NP_620591.3 3 - 4 416 143
XM_005252719.4 XP_005252776.1 3 - 4 410 143
XM_005252720.4 XP_005252777.1 3 - 4 410 143
XM_011519819.1 XP_011518121.1 3 - 4 298 143
XM_011519822.1 XP_011518124.1 3 - 4 410 143
XM_011519829.2 XP_011518131.2 3 - 4 407 143
XM_017017067.1 XP_016872556.1 3 - 4 302 143
GenBank mRNA AF125569.1 - 2 - 3 334 143
AF176070.1 - 2 - 3 349 143
AF176071.1 - 2 - 3 184 143
AK128812.1 - 4 - 5 488 143
AK298692.1 - 3 - 4 242 143
AK307638.1 - 4 - 5 488 143
AY039001.1 - 3 - 4 285 143
BC016693.1 - 3 - 4 420 143
BG506933.1 - 3 - 4 417 143

View Full Product Details