The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012393.2 | NP_001012393.1 | 5 - 6 | 922 | 70 |
NM_001319103.1 | NP_001306032.1 | 4 - 5 | 1200 | 70 | |
NM_001319104.1 | NP_001306033.1 | 4 - 5 | 837 | 70 | |
NM_001319105.1 | NP_001306034.1 | 5 - 6 | 746 | 70 | |
NM_001319106.1 | NP_001306035.1 | 4 - 5 | 1197 | 70 | |
NM_002545.4 | NP_002536.1 | 4 - 5 | 1200 | 70 | |
XM_006718846.2 | XP_006718909.1 | 5 - 6 | 1145 | 70 | |
XM_011542856.2 | XP_011541158.1 | 6 - 7 | 1458 | 70 | |
GenBank mRNA | AK289695.1 | - | 4 - 5 | 828 | 70 |
AK299528.1 | - | 6 - 7 | 595 | 70 | |
AK299908.1 | - | 5 - 6 | 709 | 70 | |
AK314077.1 | - | 4 - 5 | 1197 | 70 | |
AK315859.1 | - | 4 - 5 | 527 | 70 | |
AK316273.1 | - | 4 - 5 | 624 | 70 | |
AK316491.1 | - | 5 - 6 | 746 | 70 | |
BC074742.2 | - | 4 - 5 | 679 | 70 | |
BC074773.2 | - | 4 - 5 | 682 | 70 | |
BC117254.1 | - | 4 - 5 | 678 | 70 | |
BC126251.1 | - | 4 - 5 | 678 | 70 | |
BC143945.1 | - | 4 - 5 | 675 | 70 | |
BC143946.1 | - | 4 - 5 | 678 | 70 | |
BX537377.1 | - | 5 - 6 | 770 | 70 | |
EU562295.1 | - | 4 - 5 | 1234 | 70 | |
EU562296.1 | - | 5 - 6 | 712 | 70 | |
EU562297.1 | - | 5 - 6 | 649 | 70 | |
EU562298.1 | - | 4 - 5 | 627 | 70 | |
EU562299.1 | - | 4 - 5 | 539 | 70 | |
EU562300.1 | - | 3 - 4 | 463 | 70 | |
L34774.1 | - | 4 - 5 | 705 | 70 |