The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282424.2 | NP_001269353.1 | 7 - 8 | 933 | 72 |
NM_144670.5 | NP_653271.2 | 18 - 19 | 2418 | 72 | |
XM_011520566.2 | XP_011518868.1 | 18 - 19 | 2375 | 72 | |
XM_011520567.2 | XP_011518869.1 | 18 - 19 | 2375 | 72 | |
XM_017018868.1 | XP_016874357.1 | 18 - 19 | 2372 | 72 | |
XM_017018869.1 | XP_016874358.1 | 18 - 19 | 2372 | 72 | |
XM_017018870.1 | XP_016874359.1 | 18 - 19 | 2336 | 72 | |
GenBank mRNA | AK122624.1 | - | 18 - 19 | 2312 | 72 |
AK123591.1 | - | 1 - 2 | 71 | 72 | |
AK123592.1 | - | 7 - 8 | 888 | 72 | |
AK123601.1 | - | 15 - 16 | 1795 | 72 | |
AK290355.1 | - | 18 - 19 | 2271 | 72 | |
AK302555.1 | - | 7 - 8 | 933 | 72 | |
AL833479.1 | - | 16 - 17 | 1934 | 72 | |
BX648231.1 | - | 19 - 20 | 2270 | 72 | |
EL594621.1 | - | 3 - 4 | 72 |