Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A2ML1
Entrez Gene ID
144568
Gene Name
alpha-2-macroglobulin like 1
Gene Aliases
CPAMD9
Chromosome Location
Chr.12: 8822472 - 8887202 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282424.2 NP_001269353.1 19 - 20 2627 96
NM_144670.5 NP_653271.2 30 - 31 4112 96
XM_011520566.2 XP_011518868.1 30 - 31 4069 96
XM_011520567.2 XP_011518869.1 30 - 31 4069 96
XM_017018868.1 XP_016874357.1 30 - 31 4066 96
XM_017018869.1 XP_016874358.1 30 - 31 4066 96
XM_017018870.1 XP_016874359.1 30 - 31 4030 96
GenBank mRNA AK057908.1 - 1 - 2 114 96
AK096448.1 - 8 - 9 1101 96
AK122624.1 - 24 - 25 3198 96
AK123591.1 - 13 - 14 1765 96
AK123592.1 - 19 - 20 2582 96
AK123601.1 - 27 - 28 3489 96
AK290355.1 - 30 - 31 3965 96
AL832139.1 - 30 - 31 3963 96
AL833479.1 - 28 - 29 3628 96
BC093840.1 - 1 - 2 65 96
BC112131.1 - 1 - 2 65 96
BF330285.1 - 1 - 2 96
BX647674.1 - 12 - 13 1567 96
BX648231.1 - 31 - 32 3964 96

View Full Product Details