The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144670.5 | NP_653271.2 | 8 - 9 | 1028 | 72 |
XM_011520566.2 | XP_011518868.1 | 8 - 9 | 946 | 72 | |
XM_011520567.2 | XP_011518869.1 | 8 - 9 | 946 | 72 | |
XM_017018868.1 | XP_016874357.1 | 8 - 9 | 946 | 72 | |
XM_017018869.1 | XP_016874358.1 | 8 - 9 | 946 | 72 | |
XM_017018870.1 | XP_016874359.1 | 8 - 9 | 946 | 72 | |
GenBank mRNA | AK122624.1 | - | 8 - 9 | 922 | 72 |
AK123601.1 | - | 5 - 6 | 405 | 72 | |
AK290355.1 | - | 8 - 9 | 881 | 72 | |
AL832139.1 | - | 8 - 9 | 879 | 72 | |
AL833479.1 | - | 6 - 7 | 544 | 72 | |
BX648231.1 | - | 8 - 9 | 880 | 72 |