The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130455.1 | NP_001123927.1 | 22 - 23 | 2309 | 70 |
NM_001130976.1 | NP_001124448.1 | 21 - 22 | 2499 | 70 | |
NM_001130977.1 | NP_001124449.1 | 21 - 22 | 2499 | 70 | |
NM_001130978.1 | NP_001124450.1 | 22 - 23 | 2541 | 70 | |
NM_001130979.1 | NP_001124451.1 | 23 - 24 | 2634 | 70 | |
NM_001130980.1 | NP_001124452.1 | 22 - 23 | 2592 | 70 | |
NM_001130981.1 | NP_001124453.1 | 22 - 23 | 2592 | 70 | |
NM_001130982.1 | NP_001124454.1 | 23 - 24 | 2402 | 70 | |
NM_001130983.1 | NP_001124455.1 | 22 - 23 | 2309 | 70 | |
NM_001130984.1 | NP_001124456.1 | 21 - 22 | 2267 | 70 | |
NM_001130985.1 | NP_001124457.1 | 22 - 23 | 2360 | 70 | |
NM_001130986.1 | NP_001124458.1 | 21 - 22 | 2267 | 70 | |
NM_001130987.1 | NP_001124459.1 | 22 - 23 | 2360 | 70 | |
NM_003494.3 | NP_003485.1 | 22 - 23 | 2541 | 70 | |
XM_005264584.4 | XP_005264641.1 | 23 - 24 | 2417 | 70 | |
XM_005264585.4 | XP_005264642.1 | 23 - 24 | 3102 | 70 | |
GenBank mRNA | AF075575.1 | - | 22 - 23 | 2538 | 70 |
AK074104.1 | - | 8 - 9 | 811 | 70 | |
BC171803.1 | - | 22 - 23 | 2514 | 70 | |
DQ267935.1 | - | 22 - 23 | 2309 | 70 | |
EU515155.1 | - | 23 - 24 | 2535 | 70 | |
EU515156.1 | - | 21 - 22 | 2400 | 70 | |
EU515157.1 | - | 22 - 23 | 2442 | 70 | |
EU515158.1 | - | 22 - 23 | 2493 | 70 | |
EU515159.1 | - | 21 - 22 | 2400 | 70 | |
EU515160.1 | - | 22 - 23 | 2493 | 70 | |
EU515161.1 | - | 23 - 24 | 2402 | 70 | |
EU515162.1 | - | 21 - 22 | 2267 | 70 | |
EU515163.1 | - | 22 - 23 | 2309 | 70 | |
EU515164.1 | - | 22 - 23 | 2360 | 70 | |
EU515165.1 | - | 21 - 22 | 2267 | 70 | |
EU515166.1 | - | 22 - 23 | 2360 | 70 |