Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DYSF
Entrez Gene ID
8291
Gene Name
dysferlin
Gene Aliases
FER1L1, LGMD2B, MMD1
Chromosome Location
Chr.2: 71453155 - 71686763 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130455.1 NP_001123927.1 27 - 28 3074 81
NM_001130976.1 NP_001124448.1 26 - 27 3264 81
NM_001130977.1 NP_001124449.1 26 - 27 3264 81
NM_001130978.1 NP_001124450.1 27 - 28 3306 81
NM_001130979.1 NP_001124451.1 28 - 29 3399 81
NM_001130980.1 NP_001124452.1 27 - 28 3357 81
NM_001130981.1 NP_001124453.1 27 - 28 3357 81
NM_001130982.1 NP_001124454.1 28 - 29 3167 81
NM_001130983.1 NP_001124455.1 27 - 28 3074 81
NM_001130984.1 NP_001124456.1 26 - 27 3032 81
NM_001130985.1 NP_001124457.1 27 - 28 3125 81
NM_001130986.1 NP_001124458.1 26 - 27 3032 81
NM_001130987.1 NP_001124459.1 27 - 28 3125 81
NM_003494.3 NP_003485.1 27 - 28 3306 81
XM_005264584.4 XP_005264641.1 28 - 29 3182 81
XM_005264585.4 XP_005264642.1 28 - 29 3867 81
GenBank mRNA AF075575.1 - 27 - 28 3303 81
AK074104.1 - 13 - 14 1576 81
BC171803.1 - 27 - 28 3279 81
DB292971.1 - 4 - 5 465 81
DQ267935.1 - 27 - 28 3074 81
EU515155.1 - 28 - 29 3300 81
EU515156.1 - 26 - 27 3165 81
EU515157.1 - 27 - 28 3207 81
EU515158.1 - 27 - 28 3258 81
EU515159.1 - 26 - 27 3165 81
EU515160.1 - 27 - 28 3258 81
EU515161.1 - 28 - 29 3167 81
EU515162.1 - 26 - 27 3032 81
EU515163.1 - 27 - 28 3074 81
EU515164.1 - 27 - 28 3125 81
EU515165.1 - 26 - 27 3032 81
EU515166.1 - 27 - 28 3125 81

View Full Product Details