Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GALK2
Entrez Gene ID
2585
Gene Name
galactokinase 2
Gene Aliases
GK2
Chromosome Location
Chr.15: 49155757 - 49367883 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001556.2 NP_001001556.1 9 - 10 1377 67
NM_001289030.1 NP_001275959.1 10 - 11 1609 67
NM_001289031.1 NP_001275960.1 9 - 10 1462 67
NM_002044.3 NP_002035.1 9 - 10 1479 67
XM_005254279.3 XP_005254336.1 9 - 10 1268 67
XM_005254280.3 XP_005254337.1 9 - 10 1359 67
XM_005254284.3 XP_005254341.1 8 - 9 1260 67
XM_006720461.3 XP_006720524.1 9 - 10 1268 67
XM_006720462.3 XP_006720525.1 9 - 10 1249 67
XM_006720463.3 XP_006720526.1 8 - 9 1149 67
XM_006720464.3 XP_006720527.1 7 - 8 996 67
XM_011521441.2 XP_011519743.1 10 - 11 1472 67
XM_017022062.1 XP_016877551.1 8 - 9 1150 67
XM_017022063.1 XP_016877552.1 7 - 8 1107 67
XM_017022064.1 XP_016877553.1 7 - 8 1325 67
XM_017022065.1 XP_016877554.1 10 - 11 1524 67
XM_017022066.1 XP_016877555.1 10 - 11 1545 67
GenBank mRNA AF461816.1 - 9 - 10 1357 67
AK098049.1 - 11 - 12 1478 67
AK301361.1 - 9 - 10 1232 67
AK316440.1 - 10 - 11 1343 67
AL832745.1 - 9 - 10 1359 67
BC005141.2 - 9 - 10 1247 67
BT006901.1 - 9 - 10 1172 67
M84443.1 - 9 - 10 1192 67

View Full Product Details