The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001556.2 | NP_001001556.1 | 3 - 4 | 470 | 91 |
NM_001289030.1 | NP_001275959.1 | 4 - 5 | 702 | 91 | |
NM_001289031.1 | NP_001275960.1 | 3 - 4 | 555 | 91 | |
NM_002044.3 | NP_002035.1 | 3 - 4 | 572 | 91 | |
XM_005254279.3 | XP_005254336.1 | 3 - 4 | 361 | 91 | |
XM_005254280.3 | XP_005254337.1 | 3 - 4 | 452 | 91 | |
XM_005254284.3 | XP_005254341.1 | 3 - 4 | 452 | 91 | |
XM_006720461.3 | XP_006720524.1 | 3 - 4 | 361 | 91 | |
XM_006720462.3 | XP_006720525.1 | 3 - 4 | 342 | 91 | |
XM_006720463.3 | XP_006720526.1 | 3 - 4 | 341 | 91 | |
XM_006720464.3 | XP_006720527.1 | 3 - 4 | 341 | 91 | |
XM_017022062.1 | XP_016877551.1 | 3 - 4 | 342 | 91 | |
XM_017022063.1 | XP_016877552.1 | 3 - 4 | 452 | 91 | |
GenBank mRNA | AF461816.1 | - | 3 - 4 | 450 | 91 |
AK301361.1 | - | 3 - 4 | 325 | 91 | |
AK316440.1 | - | 4 - 5 | 436 | 91 | |
AL832745.1 | - | 3 - 4 | 449 | 91 | |
BC005141.2 | - | 3 - 4 | 340 | 91 | |
BQ921903.1 | - | 3 - 4 | 283 | 91 | |
BT006901.1 | - | 3 - 4 | 265 | 91 | |
EB386877.1 | - | 1 - 2 | 101 | 91 | |
M84443.1 | - | 3 - 4 | 285 | 91 |