Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCE1
Entrez Gene ID
6059
Gene Name
ATP binding cassette subfamily E member 1
Gene Aliases
ABC38, OABP, RLI, RLI1, RNASEL1, RNASELI, RNS4I
Chromosome Location
Chr.4: 145098004 - 145129524 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040876.1 NP_001035809.1 6 - 7 985 121
NM_002940.2 NP_002931.2 6 - 7 988 121
GenBank mRNA AB062293.1 - 6 - 7 662 121
AK315198.1 - 6 - 7 681 121
BC016283.1 - 6 - 7 650 121
BT009779.1 - 5 - 6 545 121
X76388.1 - 6 - 7 121

View Full Product Details