Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCE1
Entrez Gene ID
6059
Gene Name
ATP binding cassette subfamily E member 1
Gene Aliases
ABC38, OABP, RLI, RLI1, RNASEL1, RNASELI, RNS4I
Chromosome Location
Chr.4: 145098004 - 145129524 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040876.1 NP_001035809.1 9 - 10 1245 123
NM_002940.2 NP_002931.2 9 - 10 1248 123
GenBank mRNA AB062293.1 - 9 - 10 922 123
AK315198.1 - 9 - 10 941 123
BC016283.1 - 9 - 10 910 123
BT009779.1 - 8 - 9 805 123
X74987.1 - 3 - 4 215 123
X76388.1 - 9 - 10 922 123

View Full Product Details