The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005567.3 | NP_005558.1 | 4 - 5 | 550 | 117 |
GenBank mRNA | AK301040.1 | - | - | 458 | 117 |
AK307665.1 | - | 4 - 5 | 471 | 117 | |
BC002403.2 | - | 4 - 5 | 483 | 117 | |
BC002998.1 | - | 4 - 5 | 483 | 117 | |
BC015761.1 | - | 4 - 5 | 515 | 117 | |
KU178164.1 | - | 3 - 4 | 371 | 117 | |
L13210.1 | - | 4 - 5 | 550 | 117 | |
X79089.1 | - | 4 - 5 | 502 | 117 |