Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EYA1
Entrez Gene ID
2138
Gene Name
EYA transcriptional coactivator and phosphatase 1
Gene Aliases
BOP, BOR, BOS1, OFC1
Chromosome Location
Chr.8: 71197433 - 71548130 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000503.5 NP_000494.2 2 - 3 634 64
NM_001288574.1 NP_001275503.1 2 - 3 634 64
NM_001288575.1 NP_001275504.1 2 - 3 634 64
NM_172058.3 NP_742055.1 1 - 2 226 64
NM_172059.3 NP_742056.1 1 - 2 48 64
XM_011517483.2 XP_011515785.1 1 - 2 4934 64
XM_011517484.2 XP_011515786.2 3 - 4 392 64
XM_017013201.1 XP_016868690.1 3 - 4 420 64
XM_017013202.1 XP_016868691.1 3 - 4 264 64
XM_017013203.1 XP_016868692.1 3 - 4 392 64
XM_017013204.1 XP_016868693.1 3 - 4 393 64
XM_017013205.1 XP_016868694.1 3 - 4 392 64
XM_017013206.1 XP_016868695.1 4 - 5 373 64
XM_017013207.1 XP_016868696.1 3 - 4 391 64
XM_017013208.1 XP_016868697.1 2 - 3 659 64
XM_017013209.1 XP_016868698.1 3 - 4 281 64
XM_017013210.1 XP_016868699.1 3 - 4 393 64
XM_017013213.1 XP_016868702.1 1 - 2 5136 64
GenBank mRNA AJ000097.1 - 1 - 2 171 64
AJ000098.1 - 2 - 3 632 64
AK127798.1 - 2 - 3 134 64
AK295897.1 - 2 - 3 143 64
AK307379.1 - 2 - 3 461 64
BC121798.1 - 1 - 2 138 64
BC121799.1 - 1 - 2 138 64
DC360066.1 - 1 - 2 226 64
DC409057.1 - 1 - 2 281 64

View Full Product Details