The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000503.5 | NP_000494.2 | 6 - 7 | 1060 | 65 |
NM_172058.3 | NP_742055.1 | 5 - 6 | 652 | 65 | |
NM_172060.3 | NP_742057.1 | 4 - 5 | 524 | 65 | |
XM_011517483.2 | XP_011515785.1 | 5 - 6 | 5357 | 65 | |
XM_017013201.1 | XP_016868690.1 | 7 - 8 | 846 | 65 | |
XM_017013202.1 | XP_016868691.1 | 7 - 8 | 690 | 65 | |
XM_017013203.1 | XP_016868692.1 | 7 - 8 | 815 | 65 | |
XM_017013205.1 | XP_016868694.1 | 7 - 8 | 818 | 65 | |
XM_017013206.1 | XP_016868695.1 | 8 - 9 | 799 | 65 | |
XM_017013207.1 | XP_016868696.1 | 7 - 8 | 817 | 65 | |
XM_017013208.1 | XP_016868697.1 | 6 - 7 | 1082 | 65 | |
XM_017013211.1 | XP_016868700.1 | 6 - 7 | 690 | 65 | |
XM_017013212.1 | XP_016868701.1 | 5 - 6 | 957 | 65 | |
XM_017013213.1 | XP_016868702.1 | 5 - 6 | 5495 | 65 | |
GenBank mRNA | AJ000097.1 | - | 5 - 6 | 597 | 65 |
AJ000098.1 | - | 6 - 7 | 1058 | 65 | |
BC121798.1 | - | 4 - 5 | 883 | 65 | |
BC121799.1 | - | 5 - 6 | 564 | 65 |