The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303245.1 | NP_001290174.1 | 10 - 11 | 1057 | 130 |
NM_001303246.1 | NP_001290175.1 | 10 - 11 | 1057 | 130 | |
NM_006506.3 | NP_006497.2 | 10 - 11 | 1057 | 130 | |
XM_011513059.2 | XP_011511361.1 | 11 - 12 | 2206 | 130 | |
XM_017006968.1 | XP_016862457.1 | 11 - 12 | 1190 | 130 | |
XM_017006969.1 | XP_016862458.1 | 11 - 12 | 1189 | 130 | |
XM_017006970.1 | XP_016862459.1 | 11 - 12 | 1196 | 130 | |
XM_017006971.1 | XP_016862460.1 | 11 - 12 | 1198 | 130 | |
XM_017006972.1 | XP_016862461.1 | 9 - 10 | 921 | 130 | |
GenBank mRNA | AF115573.1 | - | 10 - 11 | 1020 | 130 |
AK292016.1 | - | 10 - 11 | 1055 | 130 | |
AK301015.1 | - | 11 - 12 | 996 | 130 | |
D78155.1 | - | 10 - 11 | 1020 | 130 | |
D82880.1 | - | 10 - 11 | 1020 | 130 |