Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASA2
Entrez Gene ID
5922
Gene Name
RAS p21 protein activator 2
Gene Aliases
GAP1M
Chromosome Location
Chr.3: 141487047 - 141615363 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303245.1 NP_001290174.1 6 - 7 642 140
NM_001303246.1 NP_001290175.1 6 - 7 642 140
NM_006506.3 NP_006497.2 6 - 7 642 140
XM_011513059.2 XP_011511361.1 7 - 8 1791 140
XM_017006968.1 XP_016862457.1 7 - 8 775 140
XM_017006969.1 XP_016862458.1 7 - 8 774 140
XM_017006970.1 XP_016862459.1 7 - 8 781 140
XM_017006971.1 XP_016862460.1 7 - 8 783 140
GenBank mRNA AF115573.1 - 6 - 7 605 140
AK292016.1 - 6 - 7 640 140
AK301015.1 - 7 - 8 581 140
D82880.1 - 6 - 7 605 140

View Full Product Details