The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006892.3 | NP_008823.1 | 20 - 21 | 2551 | 85 |
NM_175848.1 | NP_787044.1 | 19 - 20 | 2491 | 85 | |
NM_175850.2 | NP_787046.1 | 19 - 20 | 2470 | 85 | |
GenBank mRNA | AF129267.1 | - | 3 - 4 | 325 | 85 |
AF129269.1 | - | 3 - 4 | 325 | 85 | |
AF156488.1 | - | 20 - 21 | 2344 | 85 | |
AF176228.1 | - | 19 - 20 | 2470 | 85 | |
AF331857.1 | - | 20 - 21 | 2551 | 85 | |
DQ321787.1 | - | 19 - 20 | 2585 | 85 |