Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF9
Entrez Gene ID
23229
Gene Name
Cdc42 guanine nucleotide exchange factor 9
Gene Aliases
COLLYBISTIN, EIEE8, HPEM-2, PEM-2, PEM2
Chromosome Location
Chr.X: 63634967 - 63786025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173479.1 NP_001166950.1 7 - 8 1546 83
NM_001173480.1 NP_001166951.1 7 - 8 1952 83
NM_015185.2 NP_056000.1 8 - 9 2143 83
XM_005262249.2 XP_005262306.1 8 - 9 2205 83
XM_005262250.3 XP_005262307.1 8 - 9 2103 83
XM_005262251.1 XP_005262308.1 8 - 9 1453 83
XM_005262252.1 XP_005262309.1 9 - 10 1472 83
XM_011530890.1 XP_011529192.1 9 - 10 1887 83
XM_011530892.1 XP_011529194.1 8 - 9 1711 83
XM_017029363.1 XP_016884852.1 9 - 10 2232 83
XM_017029364.1 XP_016884853.1 10 - 11 2009 83
XM_017029365.1 XP_016884854.1 8 - 9 2110 83
XM_017029366.1 XP_016884855.1 9 - 10 1891 83
XM_017029367.1 XP_016884856.1 9 - 10 1880 83
XM_017029368.1 XP_016884857.1 9 - 10 2181 83
XM_017029369.1 XP_016884858.1 7 - 8 2073 83
XM_017029370.1 XP_016884859.1 7 - 8 1982 83
XM_017029371.1 XP_016884860.1 7 - 8 1971 83
XM_017029372.1 XP_016884861.1 7 - 8 1321 83
XM_017029373.1 XP_016884862.1 9 - 10 2246 83
XM_017029374.1 XP_016884863.1 6 - 7 1802 83
GenBank mRNA AB007884.1 - 8 - 9 2105 83
AK289993.1 - 8 - 9 1557 83
AK294721.1 - 3 - 4 530 83
AK295033.1 - 7 - 8 1558 83
AK295178.1 - 8 - 9 1453 83
AL832116.1 - 8 - 9 1714 83
BC117406.1 - 8 - 9 1454 83

View Full Product Details