The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017011051.1 | XP_016866540.1 | 7 - 8 | 2623 | 88 |
XM_017011052.1 | XP_016866541.1 | 7 - 8 | 2623 | 88 | |
XM_017011053.1 | XP_016866542.1 | 9 - 10 | 2799 | 88 | |
XM_017011054.1 | XP_016866543.1 | 7 - 8 | 2623 | 88 | |
GenBank mRNA | AJ277275.1 | - | 7 - 8 | 1989 | 88 |
AJ277276.1 | - | 7 - 8 | 1989 | 88 |